Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rab7a protein, rat (His, solution) | 100 UG
SDP

Catalog No. 5000291019
Encompass_Preferred

RAB7A Protein, Rat (His, solution) is a recombinant rat RAB7A small GTPase supplied as a 0.22 μm filtered protein solution with an N-terminal His tag. The material is formulated in PBS with 10% glycerol, has an approximate molecular weight of 25.7 kDa, and is intended for research use with storage at low temperatures to maintain stability.

  • Recombinant rat RAB7A with N-terminal His tag for affinity purification.
  • Supplied as a filtered solution in PBS with 10% glycerol for stability.
  • Approximate molecular weight of 25.7 kDa for experimental planning.
  • Suitable for biochemical and cell biology assays requiring purified protein.
  • Store at -80°C for long-term stability and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-91019 Supplier Medchemexpress LLC Supplier No. HYP75998A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RAB7A Protein, Rat (His, solution) is a recombinant rat RAB7A small GTPase supplied as a 0.22 μm filtered protein solution with an N-terminal His tag. The material is formulated in PBS with 10% glycerol, has an approximate molecular weight of 25.7 kDa, and is intended for research use with storage at low temperatures to maintain stability.

  • Recombinant rat RAB7A with N-terminal His tag for affinity purification.
  • Supplied as a filtered solution in PBS with 10% glycerol for stability.
  • Approximate molecular weight of 25.7 kDa for experimental planning.
  • Suitable for biochemical and cell biology assays requiring purified protein.
  • Store at -80°C for long-term stability and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.