Learn More
Aviva Systems Biology Corp RAB9A Antibody - middle region (ARP46100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB9A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAB9A. Alternate names: RAB9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-285-240