Learn More
Aviva Systems Biology Corp Rab9b Antibody - N-terminal region (ARP56895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rab9b. It was validated on Western Blot. . Alternate names: 9330195C02Rik, MGC130383ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-233