Learn More
Aviva Systems Biology Corp RAB9B Antibody - N-terminal region (ARP56894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAB9B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RAB9B. Alternate names: RAB9L, Rab-9LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-232