Learn More
Grainger Rabbeting Cut Profile Router Bit 1-1/4
Catalog No. 502766267
Encompass_Preferred
The WESTWARD Rabbeting Cut Profile Router Bit is a 1/2" shank router bit with a 1/4" cutting diameter and 1/4" rabbeting depth. It is made of solid carbide and has 2 flutes. This rabbeting router bit is designed for use on wood, plywood, and laminates. It creates a 1/4" rabbet for joining two workpieces. The bit has a cutting length of 1-1/4". It should be run at speeds between 16,000-24,000 RPM with climb or conventional cutting. Proper router speed and feed rate is important for clean cuts.
Catalog No. 50-276-6267 Supplier Grainger Supplier No. 16Y576
Add to Cart
Add to Cart
The WESTWARD Rabbeting Cut Profile Router Bit is a 1/2" shank router bit with a 1/4" cutting diameter and 1/4" rabbeting depth. It is made of solid carbide and has 2 flutes. This rabbeting router bit is designed for use on wood, plywood, and laminates. It creates a 1/4" rabbet for joining two workpieces. The bit has a cutting length of 1-1/4". It should be run at speeds between 16,000-24,000 RPM with climb or conventional cutting. Proper router speed and feed rate is important for clean cuts.