Learn More
Immunoreagents Inc RABBIT A-GOAT IGG ALP 0.5 MG
Catalog No. 50271991
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG Fc, Affinity Pure, min x w/Human Serum Proteins, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-991 Supplier Immunoreagents Inc Supplier No. RBXGT004EALP
Secondary Antibody; Rabbit anti-goat IgG Fc, Affinity Pure, min x w/Human Serum Proteins, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA