Learn More
Immunoreagents Inc RABBIT A-GOAT IGG BIO 0.5MG
Catalog No. 50270795
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-795 Supplier Immunoreagents Inc Supplier No. RBXGT003GBIO
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC