Learn More
Immunoreagents Inc RABBIT A-GOAT IGG BIO 1MG
Supplier: Immunoreagents Inc RBXGT003EBIO
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC