Learn More
Immunoreagents Inc RABBIT A-GOAT IGG DYLIGHT 800
Catalog No. 50271989
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, DyLight 800 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-989 Supplier Immunoreagents Inc Supplier No. RBXGT003G800NHSX
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, DyLight 800 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS