Learn More
Immunoreagents Inc RABBIT A-GOAT IGG FITC 0.5MG
Catalog No. 50271756
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, FITC Conjugate, Host: Rabbit, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-756 Supplier Immunoreagents Inc Supplier No. RBXGT003JFITC
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, FITC Conjugate, Host: Rabbit, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS