Learn More
Immunoreagents Inc RABBIT A-GOAT IGG FITC 1 MG
Supplier:  Immunoreagents Inc RBXGT003EFITC
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, FITC Conjugate, Host: Rabbit, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-910