Learn More
Immunoreagents Inc RABBIT A-GOAT IGG(H&L) ALP 1MG
Catalog No. 50271831
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-831 Supplier Immunoreagents Inc Supplier No. RBXGT003DALP
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA