Learn More
Immunoreagents Inc RABBIT A-GOAT IGG (H&L) AP 1MG
Supplier:  Immunoreagents Inc RBXGT004E
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG Fc, Affinity Pure, min x w/Human Serum Proteins, Host: Rabbit, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-431