Learn More
Immunoreagents Inc RABBIT A-GOAT IGG (H&L) AP 1MG
Catalog No. 50271835
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-835 Supplier Immunoreagents Inc Supplier No. RBXGT003J
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, min x w/Hu, Mu or Rt lgG/SP, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC