Learn More
Immunoreagents Inc RABBIT A-GOAT IGG(H&L) BIO 2MG
Catalog No. 50271990
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG Fc, Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-990 Supplier Immunoreagents Inc Supplier No. RBXGT004DBIO
Secondary Antibody; Rabbit anti-goat IgG Fc, Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC