Learn More
Immunoreagents Inc RABBIT A-GOAT IGG HRP 0.5MG
Supplier: Immunoreagents Inc RBXGT003GHRPX
Secondary Antibody; Rabbit anti-goat IgG (H&L), F(ab)'2 fragment, Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC