Learn More
Immunoreagents Inc RABBIT A-GOAT IGG HRP 1 MG
Catalog No. 50272111
Encompass_Preferred
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-111 Supplier Immunoreagents Inc Supplier No. RBXGT003DHRPX
Secondary Antibody; Rabbit anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC