Learn More
Immunoreagents Inc RABBIT A-HAMSTER IGG AP 1 MG
Catalog No. 50270803
Encompass_Preferred
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, min x human serum proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-803 Supplier Immunoreagents Inc Supplier No. RBXHA003E
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, min x human serum proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC