Learn More
Immunoreagents Inc RABBIT A-HAMSTER IGG BIO 1MG
Catalog No. 50272093
Encompass_Preferred
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, min x human serum proteins, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-093 Supplier Immunoreagents Inc Supplier No. RBXHA003EBIO
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, min x human serum proteins, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC