Learn More
Immunoreagents Inc RABBIT A-HAMSTER IGG HRP 1MG
Catalog No. 50272197
Encompass_Preferred
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-197 Supplier Immunoreagents Inc Supplier No. RBXHA003DHRPX
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC