Learn More
Immunoreagents Inc RABBIT A-HUMAN IGG (H&L) BIO
Catalog No. 50271853
Encompass_Preferred
Secondary Antibody; Rabbit anti-human IgG (H&L) - Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-853 Supplier Immunoreagents Inc Supplier No. RBXHU003DBIO
Secondary Antibody; Rabbit anti-human IgG (H&L) - Affinity Pure, Biotin Conjugate, Host: Rabbit, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC