Learn More
Immunoreagents Inc RABBIT A-HUMAN IGG (H&L) HRP
Catalog No. 50272113
Encompass_Preferred
Secondary Antibody; Rabbit anti-human IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-113 Supplier Immunoreagents Inc Supplier No. RBXHU003DHRPX
Secondary Antibody; Rabbit anti-human IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC