Learn More
Immunoreagents Inc RABBIT A-HUMAN IGM MU HRP
Catalog No. 50271729
Encompass_Preferred
Secondary Antibody; Rabbit anti-human IgM (mu-chain) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-729 Supplier Immunoreagents Inc Supplier No. RBXHU006DHRPX
Secondary Antibody; Rabbit anti-human IgM (mu-chain) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC