Learn More
Immunoreagents Inc RABBIT A-LLAMA IGG DYLIGHT350
Catalog No. 50271533
Encompass_Preferred
Secondary Antibody; Rabbit anti-llama IgG (H&L) - Affinity Pure, DyLight 350 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL350, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-533 Supplier Immunoreagents Inc Supplier No. RBXLL003D350NHSX
Secondary Antibody; Rabbit anti-llama IgG (H&L) - Affinity Pure, DyLight 350 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL350, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS