Learn More
Immunoreagents Inc RABBIT A-MOUSE IGG AP
Catalog No. 50271170
Encompass_Preferred
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-170 Supplier Immunoreagents Inc Supplier No. RBXMU003E
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC