Learn More
Immunoreagents Inc RABBIT A-MOUSE IGG DYLIGHT550
Catalog No. 50271172
Encompass_Preferred
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, DyLight 550 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-172 Supplier Immunoreagents Inc Supplier No. RBXMU003E550NHSX
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, DyLight 550 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS