Learn More
Immunoreagents Inc RABBIT A-MOUSE IGG FITC
Catalog No. 50271866
Encompass_Preferred
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, FITC Conjugate, Host: Rabbit, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-866 Supplier Immunoreagents Inc Supplier No. RBXMU003EFITC
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, FITC Conjugate, Host: Rabbit, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS