Learn More
Immunoreagents Inc RABBIT A-MOUSE IGG HRP
Catalog No. 50272122
Encompass_Preferred
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-122 Supplier Immunoreagents Inc Supplier No. RBXMU003DHRPX
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC