Learn More
Immunoreagents Inc RABBIT A-MOUSE IGG HRP
Supplier: Immunoreagents Inc RBXMU003EHRPX
Secondary Antibody; Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, HRP Conjugate, Host: Rabbit, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC