Learn More
Immunoreagents Inc RABBIT A-RAT IGG ALP
Catalog No. 50271698
Encompass_Preferred
Secondary Antibody: Rabbit anti-rat IgG (H&L) - Affinity Pure, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-698 Supplier Immunoreagents Inc Supplier No. RBXRT003DALP
Secondary Antibody: Rabbit anti-rat IgG (H&L) - Affinity Pure, ALP Conjugate, Host: Rabbit, Format: Liq, Label: ALP, Applications: WB, ELISA