Learn More
Immunoreagents Inc RABBIT A-RAT IGG AP
Catalog No. 50271277
Encompass_Preferred
Secondary Antibody: Rabbit anti-rat IgG (H&L) - Affinity Pure, min x w/Hu Serum Proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-277 Supplier Immunoreagents Inc Supplier No. RBXRT003E
Secondary Antibody: Rabbit anti-rat IgG (H&L) - Affinity Pure, min x w/Hu Serum Proteins, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC