Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rabbit actin native Protein from rabbit muscle

Catalog No. 89932440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-440 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-440 Supplier Abnova™ Supplier No. P5346
Add to Cart
Add to Cart

Used for AP, SDS-PAGE

Specifications

For Use With (Application) Antibody Production, SDS-PAGE
Formulation In 10mM Tris/HCl, pH 8.0, 0.2mM CaCl,
Molecular Weight (g/mol) 43kDa
Name Actin (Rabbit) Native Protein
Preparation Method Native protein purified from rabbit muscle
Quantity 100 μg
Storage Requirements Store at 4°C. This product is stable for at least 3 months. After reconstitution, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name ACTIN
Species Rabbit
Protein Tag None
Expression System Native protein purified from rabbit muscle
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.