Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit Angiopoietin-1 ELISA Kit

Catalog No. ER1RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ER1RBX5 5 x 96 Tests
ER1RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ER1RBX5 Supplier Invitrogen™ Supplier No. ER1RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Rabbit Angiopoietin-1 quantitates rabbit Angiopoietin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rabbit Angiopoietin-1.

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15389
Assay Range 0.1-25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Rabbit
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO1881110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188;
Gene ID (Entrez) 100342781, 284
Gene Symbol Angpt1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Angiopoietin 1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.