Learn More
Biomatik Corporation Rabbit Angiopoietin 2 (ANGPT2) CLIA Kit
Supplier:  Biomatik Corporation EKU11855
Encompass_Preferred
Rabbit Angiopoietin 2 (ANGPT2) CLIA Kit for detecting ANGPT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: G1T9B3. Gene ID: 100353377. Target Synonyms: ANG2; AGPT2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7476