Learn More
Biomatik Corporation Rabbit Angiotensin II (AngII) ELISA Kit
Supplier:  Biomatik Corporation EKU02408
Encompass_Preferred
Rabbit Angiotensin II (AngII) ELISA Kit for detecting AngII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 4.94-400pg/ml. Sensitivity: 1.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: G1SDD6. Gene ID: 100358371. Target Synonyms: Ang-II; Angiotensin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4973