Learn More
Biomatik Corporation Rabbit Annexin A1 (ANXA1) ELISA Kit
Supplier:  Biomatik Corporation EKU02424
Encompass_Preferred
Rabbit Annexin A1 (ANXA1) ELISA Kit for detecting ANXA1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51662. Gene ID: 100328962. Target Synonyms: ANX-A1; ANX1; LPC1; Lipocortin I; Chromobindin-9; Calpactin II; Phospholipase A2 inhibitory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6452