Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-β-Actin (High Potency), MP Biomedicals™

Catalog No. MP08L100027 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100027 Supplier MP Biomedicals, Inc Supplier No. 08L100027
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

  • Rabbit anti-β-Actin mAb (High Potency) offers high specificity and potency and consistency against antigen actins expressed by human, mouse, rat, chicken, zebrafish, pig, and Oryza sativa species, which has been experimentally validated in Western blot assay.
  • This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscular cytoskeletal actins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Actin
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Full-length recombinant protein of human β-actin
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human, Mouse, Rat, Chicken, Zebrafish
Antibody Molecular Weight Observed at 42 kDa
Form Purified
Isotype IgG
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.