Learn More
Jackson Immuno Research Labs Rabbit Anti-Alpaca Whole Serum 2ml
SDP
Catalog No. NC3806722
Encompass_Preferred
Antisera are obtained by injecting host animals with whole serum.Polyclonal antisera from immunized hosts are lipid extracted to improve clarity, salt fractionated, dialyzed against phosphate buffered saline containing sodium azide and freeze-dried. Antisera against whole serums are obtained by immunizing host animals with whole serum. Antisera against whole IgG molecules [i.e. Anti-IgG (H+L)] are recommended for bridging PAP to primary antibodies.
Catalog No. NC3806722 Supplier Jackson Immuno Research Labs Supplier No. 328001001
Antisera are obtained by injecting host animals with whole serum.Polyclonal antisera from immunized hosts are lipid extracted to improve clarity, salt fractionated, dialyzed against phosphate buffered saline containing sodium azide and freeze-dried. Antisera against whole serums are obtained by immunizing host animals with whole serum. Antisera against whole IgG molecules [i.e. Anti-IgG (H+L)] are recommended for bridging PAP to primary antibodies.