Learn More
Enzo Life Sciences Rabbit anti-biotin linker (Ready-to-use) (6 ml)
SDP
Supplier:  Enzo Life Sciences ENZ328926000
Encompass_Preferred
This rabbit anti-biotin linker is a ready to use signal amplifying reagent optimized to increase the detection sensitivity of ENZO PATHO-GENE HPV probes. Although rabbit anti-biotin linker was created to enhance ENZO probes, it will recognize any biotin associated nucleotide or macromolecule. Host: Rabbit. Source: Partially purified IgG fraction of rabbit serum. Species Reactivity: Species independent. Application: ISH (in situ hybridization). Formulation: Liquid. Ready to use in PBS containing 1% BSA and 0.1% sodium azide. Handling: Do not freeze.
Catalog No. NC1340473