Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Bovine IgG Fc Secondary Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP173565
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173565 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173565 Supplier R&D Systems Supplier No. NBP173565

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Bovine samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Host Species Rabbit
Immunogen Rabbit anti-Bovine IgG Fc fragment Secondary Antibody was produced by repeated immunization with bovine IgG F(c) fragment in rabbit.
Purification Method Affinity Purified
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Bovine IgG F(ab).
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.