Learn More
Immunology Consultants anti-Bovine IgG h+l Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 502248865
Encompass_Preferred
RGHL‑10A is an affinity‑purified rabbit antibody targeting bovine IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong, broad reactivity to bovine immunoglobulins, making it suitable for ELISA, Western blotting, and other detection workflows involving bovine antibodies or serum components. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑10A offers consistent performance and low background, supporting reliable assay development and immunodetection.
This item is not returnable. View return policy
Catalog No. 50-224-8865 Supplier Immunology Consultants Supplier No. RGHL10A
RGHL‑10A is an affinity‑purified rabbit antibody targeting bovine IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong, broad reactivity to bovine immunoglobulins, making it suitable for ELISA, Western blotting, and other detection workflows involving bovine antibodies or serum components. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑10A offers consistent performance and low background, supporting reliable assay development and immunodetection.