Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Cocktail [iFluor 488]
Catalog No. 502272024
Encompass
GenScript MonoRab™ Rabbit Anti-Camelid VHH Cocktail [iFluor 488] is specific for Camelid VHH antibody (Llama, Alpaca and Camel). It has no cross-reactivity with mouse, rat, rabbit, goat or human immunoglobulins.
This item is not returnable. View return policy
Catalog No. 50-227-2024 Supplier Genscript Corporation Supplier No. A02021200
GenScript MonoRab™ Rabbit Anti-Camelid VHH Cocktail [iFluor 488] is specific for Camelid VHH antibody (Llama, Alpaca and Camel). It has no cross-reactivity with mouse, rat, rabbit, goat or human immunoglobulins.