Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Canine IgG (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP260673
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260673 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260673 Supplier R&D Systems Supplier No. NBP260673

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Canine samples. It is validated for ELISA.

Specifications

Antigen IgG (H+L)
Applications ELISA
Classification Polyclonal
Concentration 7.56mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2500, ELISA 1:2000-1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Symbols IGHG1
Host Species Rabbit
Immunogen Dog IgG whole molecule
Purification Method Affinity Purified
Quantity 10 mg
Primary or Secondary Secondary
Test Specificity This product was prepared from monospecific antiserum by immunoaffinity chromatography using Dog IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities.
Target Species Canine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.