Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Chicken IgG, Whole Molecule Polyclonal Secondary Antibody, FITC, MP Biomedicals

Catalog No. MP0855307 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855307 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855307 Supplier MP Biomedicals, Inc Supplier No. 0855307
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host Species: Rabbit
Species Reactivity: Ck

Immunostaining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Chicken IgG
Classification Polyclonal
Conjugate FITC
Host Species Rabbit
Quantity 2 mL
Primary or Secondary Secondary
Target Species Chicken
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.