Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit Anti-Chicken IgY Antibody (H&L) Antibody (Rhodamine), ProSci
SDP

Catalog No. 16838640 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-838-640 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-838-640 Supplier ProSci LLC Supplier No. XG6183RH0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Control Antibody

Rabbit Anti-Chicken IgY Antibody specifically detects Rabbit Anti-Chicken IgY in Chicken samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Rabbit Anti-Chicken IgY
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate Rhodamine
Dilution This antibody can be used at 1:50-1:200 for most IHC/ICC applications, with1-5% rabbit serum in dilution buffer.
Formulation Buffer containing 0.01M sodium phosphate, 0.25M sodium chloride, pH 7.6 and 25% glycerol.
Host Species Rabbit
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This antibody reacts with both heavy and light chains of chicken IgY, minimum cross-reactivity with IgG of mammalian species.
Target Species Chicken
Content And Storage Store 2 - 8 °C as an undiluted liquid for several weeks. After dilution, do not use for more than one day. -20 °C for up to one year.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.