Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit Anti-Chicken IgY Antibody (HRP), ProSci
SDP

Catalog No. 16817861 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-817-861 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-817-861 Supplier ProSci LLC Supplier No. XR91201500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Control Antibody

Rabbit Anti-Chicken IgY Antibody specifically detects Rabbit Anti-Chicken IgY in Chicken samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Specifications

Antigen Rabbit Anti-Chicken IgY
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Dilution Peroxidase Conjugated Affinity Purified anti-Chicken IgY is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency. Recommended dilutions are as follows, ELISA 1:35,000 - 1:150,000, Western Blot 1:3,500 - 1:15,000, Immunohistochemistry 1:1,000 - 1:5,000. (Optimal dilution should be determined by user).
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2; 10 mg/mL Bovine Serum Albumin (BSA); 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!
Host Species Rabbit
Immunogen Chicken IgY - whole molecule
Purification Method Peroxidase Conjugated Affinity Purified anti-Chicken IgY was prepared from monospecific antiserum by immunoaffinity chromatography using Chicken IgY coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum, Chicken IgY and Chicken Serum.
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Peroxidase Conjugated Affinity Purified anti-Chicken IgY can be stored vial at 4 °C prior to restoration. For extended storage aliquot contents and freeze at -20 °C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
Product Type Antibody
Form Lyophilized
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.