Learn More
Description
Specifications
Specifications
| Antigen | Rabbit Anti-Chicken IgY (H&L) |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Alkaline Phosphatase Conjugated Affinity Purified anti-Chicken IgY is suitable for immunoblotting (western or dot blot), ELISA and immunohistochemistry as well as other phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency. This product has been assayed against 1.0 mg of Chicken IgY in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature. A working dilution of 1:3,000 to 1:12,000 is suggested for this product. (Optimal dilution should be determined by user). |
| Formulation | 0.05M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0; 0.1% (w/v) Sodium Azide; 10 mg/mL Bovine Serum Albumin (BSA) |
| Host Species | Rabbit |
| Purification Method | Alkaline Phosphatase Conjugated Affinity Purified anti-Chicken IgY was prepared from monospecific antiserum by immunoaffinity chromatography using Chicken IgY coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Alkaline Phosphatase (calf intestine), anti-Rabbit Serum, Chicken IgY and Chicken Serum. |
| Quantity | 1.0 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
