Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody, AP

Catalog No. SA19514
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA19514 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA19514 Supplier Invitrogen™ Supplier No. SA19514
Add to Cart
Add to Cart

Rabbit Polyclonal Secondary Antibody

This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9514 has minimum cross-reactivity with IgG of mammalian species. SA1-9514 immunogen is immunoglobulin isolated from chicken serum. Store at 4°C as an undiluted liquid for several weeks. After dilution, do not use for more than one day. For long-term storage, use -20°C.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chicken IgY (H+L)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.667 mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation TBS with 25% glycerol, 10mg/mL BSA and 0.02% sodium azide; pH 8.0
Host Species Rabbit
Immunogen Immunoglobulin isolated from chicken serum.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.