Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NB7297
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7297 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7297 Supplier R&D Systems Supplier No. NB7297

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgY (H+L) Polyclonal specifically detects IgY (H+L) in Chicken samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgY (H+L)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, ELISA, Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin
Host Species Rabbit
Immunogen Chicken IgG (IgY)-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with chicken IgG and with light chains common to other chicken immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species.
Target Species Chicken
Content And Storage Store at 4C. Do not freeze.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.