Learn More
Immunology Consultants anti-Dog CRP Antibody (Rabbit) - Antiserum
SDP
Catalog No. 502248808
Encompass_Preferred
RCRP‑40 is a rabbit antiserum containing polyclonal antibodies specific for canine C‑reactive protein (CRP), a key biomarker of inflammation in veterinary diagnostics. As a whole‑serum preparation, it provides broad epitope recognition and strong signal generation across multiple assay formats. This antibody is well suited for ELISA development, immunodetection, and research applications focused on canine inflammatory responses. Produced by Immunology Consultants and supplied in a 1 mL format, RCRP‑40 offers a reliable, cost‑effective reagent for laboratories studying CRP expression in dogs or developing CRP‑based diagnostic tools.
This item is not returnable. View return policy
Catalog No. 50-224-8808 Supplier Immunology Consultants Supplier No. RCRP40
RCRP‑40 is a rabbit antiserum containing polyclonal antibodies specific for canine C‑reactive protein (CRP), a key biomarker of inflammation in veterinary diagnostics. As a whole‑serum preparation, it provides broad epitope recognition and strong signal generation across multiple assay formats. This antibody is well suited for ELISA development, immunodetection, and research applications focused on canine inflammatory responses. Produced by Immunology Consultants and supplied in a 1 mL format, RCRP‑40 offers a reliable, cost‑effective reagent for laboratories studying CRP expression in dogs or developing CRP‑based diagnostic tools.